Learn More
Gibco™ Fetal Bovine Serum, MaxSpec
Gibco MaxSpec Fetal Bovine Serum (FBS) is our highest quality FBS, meeting selective specifications to help provide consistent, reproducible results.
Brand: Gibco™ A4766801
671.64 GBP valid until 2025-12-31
Use promo code "24268" to get your promotional price.
Description
Gibco MaxSpec Fetal Bovine Serum (FBS) is our highest quality FBS, meeting selective specifications to help provide consistent, reproducible results. Only MaxSpec FBS delivers our best quality specifications and testing levels.
Gibco sera: trusted quality for consistent performance
MaxSpec Sera Features
- Use with a broad range of cell types, especially sensitive cell lines
- Our most characterized sera with the lowest endotoxin and hemoglobin levels
- Endotoxin level: ≤1 EU/mL
- Hemoglobin level: ≤15 mg/dL
- Origin: United States, Australia, or New Zealand (refer to CoA for specific origin)
Additional features of this serum include:
- Meets USP/EP guidelines with up to 96 harmonized quality specification tests
- Additional proliferation tests on top six cell lines
- EMA virus testing upon request
- Lowest risk of BSE and lower viral risk
- ISO 13485-certified; processed in FDA-registered facilities
- Triple-filtered at 0.1 μm
- Gibco bottle is easy to use in a hood, helps reduce risk of contamination, and allows you to perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Specifications
Meets USP/EP guidelines; ISO 13485 Certified | |
Store at or below -10°C. | |
Use for a broad range of cell types, especially sensitive cell lines | |
Refer to CoA for specific origin | |
Bottle | |
500 mL | |
Triple-filtered, 0.1 μm | |
≤1 EU/mL | |
≤15 mg/dL | |
Frozen |
Specialty | |
Fetal | |
Cattle/Bovine | |
Liquid | |
Fetal Bovine Serum | |
Sterile | |
Meets USP/EP guidelines with up to 96 harmonized quality specification tests; Additional proliferation tests on top 6 cell lines; EMA virus testing upon request | |
Less waste | |
Premium Plus |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2481746RP | Certificate of Analysis | 29/01/2025 | A4766801 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.