Learn More
Gibco™ Fetal Bovine Serum, qualified, Australia
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Brand: Gibco™ 10099141
667.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera category: Premium (Secure)
- Origin: Australia.
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤30 mg/dL (levels routinely ≤25 mg/dL)
Aliquot-free 50 mL bottle
The Gibco One Shot 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.
Features include:
- Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
- Engineered to freeze/thaw without splitting or breaking
- Fits in test tube racks and frees up freezer space
- No labeling mishaps with our easy-to-read package label containing all of the information you need
- 33% less waste generated than aliquoting from 500 mL bottles
All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
≤5 EU/mL | |
≤30 mg/dL | |
Frozen |
Fetal | |
Australia | |
Bottle | |
500 mL | |
Sterile | |
Sustainable packaging | |
Qualified |