missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Fetal Bovine Serum, qualified, Brazil
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C results.
£78.25 - £367.00
Specifications
Additional Assays | Basic cell culture, specialty research, and specific assays |
---|---|
Certifications/Compliance | EU-Approved |
Content And Storage | Store in freezer (–5 to –30°C) |
Country of Origin | E.U. Approved (South American) |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11573397
|
Gibco™
10270106 |
500 mL |
£367.00
500mL |
Estimated Shipment: 29-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11563397
|
Gibco™
10270098 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Sera Category: StandardOrigin: E.U. Approved (South American)
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C results.
Specifications
Basic cell culture, specialty research, and specific assays | |
Store in freezer (–5 to –30°C) | |
Liquid | |
Fetal Bovine Serum | |
Cattle/Bovine | |
Liquid |
EU-Approved | |
E.U. Approved (South American) | |
Bottle | |
Sterile-filtered | |
Virus, Mycoplasma | |
Basic cell culture, specialty research, and specific assays |