missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, E.U.-approved, South America Origin
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C. results.
£80.50 - £393.00
Specifications
Additional Assays | Basic cell culture, specialty research, and specific assays |
---|---|
Certifications/Compliance | EU-Approved |
Content And Storage | Store in freezer (–5 to –30°C) |
Country of Origin | E.U. Approved (South American) |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11550356
|
Gibco™
10500064 |
500 mL |
£393.00
500mL |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11543407
|
Gibco™
10500056 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Sera Category: StandardOrigin: E.U. Approved (South American)
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C. results.
Specifications
Basic cell culture, specialty research, and specific assays | |
Store in freezer (–5 to –30°C) | |
Liquid | |
Cattle/Bovine | |
Virus, Mycoplasma | |
Basic cell culture, specialty research, and specific assays | |
Qualified |
EU-Approved | |
E.U. Approved (South American) | |
Bottle | |
Qualified | |
Liquid | |
Heat-Inactivated |