missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, United States

Important Update: We've recently introduced Premium Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified US origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Brand:  Gibco™ 16140071

Code : 50

Additional Details : CAS Number : 64742-49-0 Weight : 0.59000kg

 View more versions of this product

Product Code. 11580516

  • £469.00 / 500mL
Estimated Shipment: 15-08-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Value Plus (Performance)
  • Origin: United States.
  • Endotoxin level: ≤10 EU/mL
  • Hemoglobin level: ≤25 mg/dL
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Fetal Bovine Serum
Heat-inactivated
Sustainable packaging
Qualified
Fetal
United States
Bottle
500 mL
≤10 EU/mL
≤25 mg/dL
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers