Learn More
Gibco™ Fetal Bovine Serum, Tet system approved, USDA-approved regions
Gibco Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems. Cell-based assays that detect the presence of tetracycline and tetracycline analogs are used to select FBS that delivers the highest range of induction.
£90.25 - £716.00
Specifications
Certifications/Compliance | ISO 13485 Certified |
---|---|
Classification | Value |
Content And Storage | Store at <-10°C. |
Species | Cattle/Bovine |
Cell Type | Use for a broad range of cell types, especially sensitive cell lines |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
16866796
|
Gibco™
A4736301 |
50 mL | One Shot |
£90.25
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16876796
|
Gibco™
A4736401 |
500 mL | Bottle |
£716.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems. Cell-based assays that detect the presence of tetracycline and tetracycline analogs are used to select FBS that delivers the highest range of induction.
- Designed to ensure the absence of detectable tetracycline and its derivatives that may interfere with accurate regulation of gene expression
- Functional testing provides confidence that unintended tetracyline gene expression will not result
- Sera Category: Specialty
Specifications
ISO 13485 Certified | |
Store at <-10°C. | |
Use for a broad range of cell types, especially sensitive cell lines | |
Mexico | |
Fetal Bovine Serum | |
Triple-filtered, 0.1 μm | |
Sustainable packaging | |
Value |
Value | |
Cattle/Bovine | |
Fetal | |
Tet-System Approved | |
Sterile | |
<50 EU/mL (levels routinely <10 EU/mL) | |
<25 mg/dL | |
Frozen |
Safety and Handling
missing translation for 'shelfLife' : 3 Years
Your input is important to us. Please complete this form to provide feedback related to the content on this product.