Learn More
Molecular Probes™ Fetal Hemoglobin Control Kit (FETALtrol™)
FETALtrol™ is a tri-level, assayed human blood control designed to help document and monitor values obtained from test methods used to determine fetal RBCs in maternal blood samples.
Brand: Molecular Probes™ FH101
Code : 50
Additional Details : Weight : 0.06000kg
Description
FETALtrol™ is a tri-level, assayed human blood control designed to help document and monitor values obtained from test methods used to determine fetal RBCs in maternal blood samples. It monitors preparation techniques, stains, reagents, and methods of data analysis for quantitative and qualitative tests. FETALtrol™ is reagent comprising D-antigen (Rho) negative adult human erythrocytes supplemented with D-antigen (Rho) positive human cord blood erythrocytes.
Application
The laboratory determination of the level of fetal cells in maternal circulation remains an important support in the obstetrical management of women with suspected uterine trauma and in the proper dose administration of Rh immune globulin. The Trillium FETALtrol™ kit provides a means of monitoring preparation techniques, stains, reagents, and methods of data analysis for quantitative and qualitative tests.
Additional Information
Use the Spectraviewer to help you plan your experiments. Additional general information about Life Technologies flow cytometry products and support is also available.
Specifications
Store in refrigerator (2°C to 8°C). | |
Microscope (KBB assay-compatible), Flow Cytometer | |
Flow Cytometry, Immunohistochemistry |
Tube | |
Fetal Hemoglobin Control Kit | |
50 Tests |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.