Learn More
FFP2 Disposable Respirator Face Mask
FFP2 Disposable respirator to protect against inhalation of droplets and particles. Available with head band and without exhalation valve.
Brand:  Roesner Mautby Meditrade ZH3161
Product Code. 17176314
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Alert:
Compliance
This product complies with the essential requirements for health and safety as they are provided by the European Regulation (EU) 2016/425 for Personal Protective Equipment.
Description
Description
  • Tested and certified FFP2 disposable respirator
  • Without exhalation valve
  • Head bands enable a more secure, tighter and comfortable fit
  • Head bands make wearing glasses more comfortable and enable good compatibility with various eye protection products
Specifications
Specifications
CE Approved, EN 149:2001+A1:2009 FFP2 NR
Single-use
Face Piece: Polypropylene and Polyester; Head Band: Polyurethane and Polyester; Nosepiece: Metal
One Size
Non-sterile
Flat-fold
FFP2 NR
Particulate Respirator with Double Head Bands
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
FFP2 Disposable Respirator Face Mask > Non-valved; Head Straps