missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Source™ FFP2 Disposable Respirator Face Mask
FFP2 Disposable respirator to protect against inhalation of droplets and particles. Available with or without exhalation valve.
£1.00
Specifications
Certifications/Compliance | CE Approved, EN 149:2001+A1:2009 FFP2 NR D |
---|---|
Color | White |
Design Type | Cup/Molded |
Disposable | Single-use |
Material | Anti-Bacterial Cotton, Non-Woven Fabric |
Product Code | Brand | With Exhalation Valve | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | With Exhalation Valve | Price | Quantity & Availability | |||||
16635042
|
New Source™
HY8620 |
No |
£1.00
Pack of 20 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
16669272
|
New Source™
HY8622 |
Yes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Polypropylene outer layers
- Cup shaped
- 16669272 has exhalation valve for increased comfort
- 16635042 is without valve
- Compatible with goggles/glasses and reduces fogging due to contoured design
- Adjustable soft-foam nosepiece ensures custom fit to increase wearer's comfort
- Seal avoids open layer around the edges
Specifications
CE Approved, EN 149:2001+A1:2009 FFP2 NR D | |
Cup/Molded | |
Anti-Bacterial Cotton, Non-Woven Fabric | |
One Size | |
Protection Against Dusts and Mists in a Wide Variety of Industrial Applications and Other Hazardous Environments | |
Non-sterile |
White | |
Single-use | |
Particulate Respirator with Double Head Bands | |
FFP2 | |
Yes |