Learn More
Coverguard™ FFP2 NR D Face Mask
The Coverguard™ FFP2 masks are single-use and protect against non-toxic solid and liquid aerosols in applications such as farming, silviculture and fishing, light industries, maintenance, infrastructure and building, DIY, as well as general use in the lab or field.
Brand:  Coverguard™ 23205
 View more versions of this product
Product Code. 12083186
  • £633.00 / Pack of 240
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PPE Cat. III FFP2 NR D
  • Disposable
  • Electrostatic filter media
  • Sliding elastic band for better adjustment
  • Latex-free, non-woven polypropylene
  • Adjustable nose clip
Specifications
Specifications
CE PPE Category III FFP2 NR D, EN 149:2001 + A1:2009
Single-use
Chemical and Pharmaceutical Industries, Light Industries, Heavy and Process Industries, Construction Finishings, Building Industry, Infrastructure
Non-Toxic Solid and Liquid Aerosols
Half Mask with Exhalation Valve
FFP2 Disposable Half Mask, Sealed Outer Contour, Exhalation Valve
Yes
Adjustable Nose Clip
Non-sterile