missing translation for 'onlineSavingsMsg'
Learn More

FG 7142, Tocris Bioscience™

Product Code. 30001100
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Benzodiazepine inverse agonist

FG 7142 is a benzodiazepine inverse agonist and anxiogenic agent. Increases tyrosine hydroxylation and causes upregulation of β-adrenoceptors in mouse cerebral cortex.

Specifications

CAS 78538-74-6
Molecular Formula C13H11N3O
MDL Number MFCD00055075
Quantity 100 mg
Synonym n-methyl-beta-carboline-3-carboxamide, unii-60po70n1bp, n-methyl-9h-pyrido 3,4-b indole-3-carboxamide, beta-carboline-3-carboxylic acid n-methylamide, 9h-pyrido 3,4-b indole-3-carboxamide, n-methyl, beta-cca methylamide, n-methyl-9h-, a-carboline-3-carboxamide, tocris-0554
InChI Key QMCOPDWHWYSJSA-UHFFFAOYSA-N
SMILES CNC(=O)C1=CC2=C(NC3=C2C=CC=C3)C=N1
IUPAC Name N-methyl-9H-pyrido[3,4-b]indole-3-carboxamide
Molecular Weight (g/mol) 225.25
PubChem CID 4375
Formula Weight 225.25
Percent Purity >99%
Chemical Name or Material FG 7142
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'