Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optics Lightguide Adapter
GREENER_CHOICE
This Fiber Optics Lightguide Adapter can be used to mount fiber optic lightguides from Volpi/Photonic on the SCHOTT Coldlightsource (KL1500HAL).
Brand:  SCHOTT AG Lighting and Imaging™ 158320
Product Code. 30052752
  • £148.00 / Each
Explore more special offers
Description
Description
  • Lightguides from Volpi or Photonic have a different endtip diameter than the SCHOTT Fiber Optic Lightguide
  • With this adapter, Lightguides from Volpi or Photonic can be used with a SCHOTT Cold Light Source
  • For use only with SCHOTT Cold Light Source (KL1500HAL)
Specifications
Specifications
Mount Adapter
KL 1500 HAL Cold Light Source