Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters
Click to view available options
Capacity:
10 mL Vacutainer Tube
100 mL Oak Ridge Tube
15 mL Conical Tube
16 mL Oak Ridge Tube
3 mL Vacutainer Tube
30 mL Oak Ridge Tube
Unit Size:
Pack of 2
Description
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support Ridge™, Vacutainer™ and conical tubes.
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.
Specifications
Specifications
| Type | Rotor Tube Adapter |
| Capacity | 15 mL Conical Tube |
| Vessels per Adapter | 5 |