missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250y Fixed Angle Rotor
Process large volumes with the six-place, 250 mL fixed angle rotor.
£12295.00
Specifications
Description | Fiberlite F14-6 x 250y Fixed Angle Rotor |
---|---|
Material | Carbon Fiber Composite |
Type | Fixed Angle Rotor |
Capacity | 6 x 250 mL |
Max. RCF | 30,100 x g |
Product Code | Brand | For Use With (Equipment) | Capacity | Max. Speed | Max. RCF | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Capacity | Max. Speed | Max. RCF | Price | Quantity & Availability | ||
10610491
|
Thermo Scientific™
78500 |
Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges | 6 x 250 mL | 14,000 rpm | 30,100 x g |
£12295.00
Each |
Estimated Shipment: 13-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Process large volumes with the six-place, 250mL Thermo Scientific™ Fiberlite™ F14-6x250y Fixed Angle Rotor.
Fiberlite rotors:- Run times are reduced with higher G-forces achieved through lightweight carbon fiber rotors.
- Corrosion-and fatigue-resistant design ensures structural integrity.
- Rotor is backed by a 15-year warranty.
Compatible with:
Thermo Scientific Sorvall™ RC-6 Plus, Sorvall Evolution™ RC Series, Thermo Scientific Jouan KR22i, Beckman™ J2 Series, Beckman J2, Avanti Series, Hitachi™ Himac CR21/CR22 Series, Nalgene™ bottles and tubes
Recommended for:
Processing lysates, homogenates, ammonium sulphate precipitation of proteins and enzymes and plant tissue homogenates.
†Performance may vary by centrifuge model.
Specifications
Fiberlite F14-6 x 250y Fixed Angle Rotor | |
Fixed Angle Rotor | |
30,100 x g | |
137 mm | |
1690 | |
62 mm | |
23° | |
Yes |
Carbon Fiber Composite | |
6 x 250 mL | |
14,000 rpm | |
37 mm | |
135 mm | |
15.5 lb. (7.03 kg) | |
8,122 x g |