Description
Combine high g-force and high capacity for 1.5mL microtubes with the Thermo Scientific™ Fiberlite™ F23-48 x 1.5mL Fixed-Angle Rotor.
Features:- Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal
- Corrosion and fatigue-resistance secures the rotor's structural integrity for unequalled durability
- Rotor is backed by a 15-year warranty
- Secure, push-button rotor exchange in less than three seconds
- Improved safety and convenience
- No tools required
- Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
- Immediate identification of a rotor
- Rotor specifications automatically loaded into centrifuge parameters
- Shortens run set-up time, improves safety
Speed Handle:
- Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
- Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
Fiberlite F23-48x1.5 Fixed Angle Rotor | |
Fixed Angle Rotor | |
57,368 x g | |
97 mm | |
137 | |
11 mm | |
Sorvall LYNX 6000 and 4000 Superspeed Centrifuges |
Carbon Fiber Composite | |
48 x 1.5 mL | |
23,000 rpm | |
64 mm | |
40 mm | |
10.0 lb. (4.54 kg) | |
45° |