Learn More
Thermo Scientific™ Fiberlite F37L-8 x 100 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
GREENER_CHOICE
Reduce the required number of runs for high-volume separations with this ultraspeed carbon fiber rotor that features two additional tube cavities.
Brand:  Thermo Scientific™ 096-087056
15043.12 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 15315252
  • £19664.13 / Each
Explore more special offers
Description
Description
Reduce the required number of runs for high-volume separations with the Thermo Scientific™ Fiberlite™ F37L-8 x 100 Fixed-Angle Rotor, an ultraspeed carbon fiber rotor that offers two additional tube cavities for an eight-place total capacity.
Fiberlite rotors:
  • Reduce run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering information:
Tube adapters are sold separately accommodating a variety of tubes in different capacities. Adapters available in packages of two. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ WX Ultracentrifuge Series and Beckman™ Floor Model L Series (except Optima) centrifuges, Nalgene™ bottles and tubes
Recommended for:
Separations of subcellular organelles or concentration of viruses.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fiberlite F37L-8 x 100 Fixed-Angle Rotor
Fixed Angle Rotor
182,460 x g
119 mm
168
38 mm
Thermo Scientific™ Sorvall™ WX ultraCentrifuges
73,600 x g
Carbon Fiber Composite
8 x 100 mL
37,000 rpm
48.8 mm
102 mm
25 lb. (11.34 kg)
25°
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Fiberlite F37L-8 x 100 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges > F37L-8x100 Fixed-Angle Rotor