Learn More
Epredia™ Fibrinogen Antibody
Perform rapid and inexpensive assessment of tissues with Epredia™ Fibrinogen, FITC Labeled Antibody, designed specifically for immunofluorescence staining.
Brand: Epredia™ RB-1924-R2
Description
Host Species: Rabbit
Species Reactivity: Reacts with fibrinogen plasma protein including fibrinogen fragments D, E, X and Y.
Immunogen: Fibrinogen isolated from human serum
Positive Control: Lymph node, spleen, liver, kidney or skin fixed and processed in the same manner as the test specimen
Cellular Localization: Lymphatic tissues, liver or in various types of diseased skin or kidney tissue
Recommended for:
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Fibrinogen | |
Polyclonal | |
FITC | |
Fibrinogen isolated from human serum | |
IVD | |
Primary | |
Prediluted |
Immunofluorescence | |
Polyclonal | |
Rabbit | |
2 mL | |
Cancer and Tumor Biology | |
Human |