Learn More
Invitrogen™ Human Fibrinogen, Native Protein
Native Protein
Brand: Invitrogen™ RP43142
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
Fibrinogen (human) Native Protein, RP-43142, from Invitrogen.
Fibrinogen, the precursor of fibrin, is the coagulable protein in the blood plasma of vertebrates. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. Fibrinogen consists as a dimer of 3 pairs of non-identical chains FGA, FGB, and FGG that are cross-linked by disulfide bonds in their N-terminal segments. The molecule has 2 terminal D domains and one central E domain, all three domains are separated when fibrinogen is degradated by plasmin. Mutations in this gene lead to several disorders, including dysfibrinogenemia, afibrinogenemia, and renal amyloidosis. RP-43142 contains native human Fibrinogen protein. RP-43142 may be used as a positive control in many immunological assays. The RP-43142 protein molecular weight is ∽340 kDa. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!Specifications
P02671 | |
Control | |
2243 | |
7.3 | |
Human | |
RUO | |
FGA | |
Unconjugated | |
Human | |
Liquid | |
Affinity chromatography |
7.8mg/mL | |
0.01M citric acid, 0.01M sodium phosphate with no preservative; pH 7.3 | |
Human Fibrinogen | |
2mg | |
-80°C, Avoid Freeze/Thaw Cycles | |
Human Fibrinogen | |
Human | |
Hu | |
Native | |
Native |