Learn More
Lonza™ Fibroblast Growth Medium
Product Code. 13419139
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. 13419139
Brand: Lonza™ CC4526
Specifications
Product Type Growth Medium
Quantity 1 Kit
Content And Storage Kit which contains a 500mL bottle of FBMTM, (CC-3131) and FGMTM-3 singlequotsTM (CC-4525-supplements and growth factors (hFGF-B, insulin, FBS and gentamicin/ amphotericin-B)
compliance-icons
Product Identifier
  • FGM-3 Bullet Kit
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Gentamicin, sulfate(salt)