missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibromodulin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526250
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology.
Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix.Specifications
Fibromodulin | |
Polyclonal | |
Unconjugated | |
FMOD | |
AI131919; AU041740; Collagen-binding 59 kDa protein; fibromodulin; FM; FMOD; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin; SLRR2E | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
14264, 2331 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P50608, Q06828 | |
FMOD | |
KLH conjugated synthetic peptide between 337-364 amino acids from the C-terminal region of human FMOD | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |