Learn More
Thermo Scientific Chemicals Fibronectin-Binding Protein
For research use
Brand: Thermo Scientific Chemicals J66903.M¬
Code : NEW
Additional Details : CAS Number : 119977-20-7 Weight : 0.00001kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Lyophilized powder | |
-30° to -10°C | |
Fibronectin-Binding Protein | |
Solid |
1mg | |
RUO | |
Native |
RUO – Research Use Only