Learn More
Gibco™ Fibronectin Human Protein, Plasma
Fibronectin Human Protein, Plasma
Brand:  Gibco™ 33016015
Product Code. 11564466
  • £1288.00 / 5mg
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Fibronectin is a large, cell surface and plasma protein that exhibits structural and adhesive properties in cell-associated fibrillar matrices. Fibronectin is one of the primary cell adhesion molecules. Human Plasma Fibronectin is isolated from human plasma by affinity chromtography. Fibronectin has a molecular weight of 440,000 and is made up of two 220,000 dalton polypeptides. Human Fibronectin is filtered (0,2UI) and supplied lyophilized.
Performance and Quality Testing
The purity is determined by SDS-PAGE. Bovine and human fibronectins are assayed for their ability to support cell attachment and spreading, and screened to confirm absence of mycoplasma or bacterial contamination.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Culture
Fibronectin Human, Plasma
Store in refrigerator (2–8°C).
Lyophilized
Extracellular Matrix & Cell Adhesion, Stem Cell Research
5 mg
Human
Room Temperature
Fibronectin