missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Physical Form | Liquid |
Melting Point | 3°C |
Boiling Point | 280°C |
Packaging | Plastic Coated Glass bottle |
Color | Brown to Yellow |
Quantity | 2.5 L |
MDL Number | 283231 |
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Safety and Handling


Product Identifier
- Ficodox
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains A§. May produce an allergic reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ficodox™, Pure, Mixed Cod Reagent, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction