missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Poly Bottle
Description
Ficoll 400 (White to Off-White Powder/Molecular Biology)
Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Physical Form | Powder/Solid |
Quantity | 500 g |
Formula Weight | ≈400,000 |
Grade | Molecular Biology |
Packaging | Poly Bottle |
Specific Rotation | Inclusive between 50° (+ or -) |
Chemical Name or Material | Ficoll™ 400 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'