missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ficoll PM400

Product Code. 11570724 Shop All Cytiva Products
Click to view available options
Quantity:
100 g
500 g
5 kg
Unit Size:
100g
500g
5kg
This item is not returnable. View return policy

Product Code. 11570724

Brand: Cytiva 17030005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. Cytiva™ Ficoll PM400 is a synthetic neutral, highly-branched hydrophilic polymer of sucrose, used for producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity.

  • For producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity
  • Neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution
  • Concentrations of up to 50% (w/v) covering densities of up to 1.2g/mL can be achieved
  • Has better osmotic properties than sucrose
  • Preserves functional and morphological integrity
  • Useful for separating cells that are sensitive to centrifugation and for separating cells of similar density but different size
  • Does not penetrate biological membranes
  • Serves as the raw material for preparation of Ficoll-Paque gradients
  • Used in other applications such as electrophoresis, hybridization, cryopreservation, and as a hapten carrier
  • Supplied as dry powders

  • Used in the formation of density gradients for the separation and isolation of eukaryotic cells, organelles, and bacterial cells, as a stabilizing agent and as a separation medium for the isolation of lymphocytes
  • Used in defined culture media, nucleic acid hybridization, electrophoresis, and immunological studies
  • Used for gradient centrifugation in all types of centrifuge rotors and for separation at unit gravity

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 kg
Content And Storage 4–30°C
For Use With (Application) Cell Isolation
Viscosity 5 ηr at 20°C: 10% (w/v), 20 ηr at 20°C: 20% (w/v), 60 ηr at 20°C : 30% (w/v, 180 ηr at 20°C : 40% (w/v) , 600 ηr at 20°C : 50% (w/v)
Endotoxin Level 0.12 EU/mg
Color White
Product Type Centrifugation Media
Density 1.2 g/mL
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.