Learn More
Ohaus™ Field Test Scale
The Field Test Scale provides accurate weighing of a wide variety of heavy materials, even under the most adverse conditions. The Field Test Scale is ruggedly built and designed for field applications where accurate measurements are essential
Brand:  Ohaus™ 80000008
Product Code. 10531222
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
15.35 in.
8.22 in.
Epoxy coated steel pan
Metal Base and Beam, Oversized Stainless Steel Pan
194.00 mm
Field Test
Mechanical Scale, 2400-11
Sliding Poise and Graduated Beam
390.00 mm
209.000 mm
2400-11
7.64 in.
In a Laboratory, in an Intrinsically Safe Environm
5.000000 g
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Spanish (MX)
Field Test Scale