missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Filling Tubes for BRAND Seripettor, Dispensette S, Dispensette III and Dispensette Organic Bottletop Dispensers

Filling tubes for all BRAND Seripettor and Dispensette models made after 1997

Brand:  BRAND™ 708216

 View more versions of this product

Product Code. 15428023

  • £23.10 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 704536 - autoclavable filling tube for seripettor, 250mm, each
  • 704538 - autoclavable filling tube for seripettor, 500mm, each
  • Telescoping filling tubes adjust to correct height without cutting.
  • Unique design allows aspiration of liquid from bottom of bottle while minimizing aspiration of precipitates.
  • Two different diameter tubes: One for 1mL, 2mL, 5mL and 10mL nominal capacity dispensers; and one for 25mL, 50mL and 100mL nominal capacity dispensers
  • Five different lengths: 70-140mm; 125-240mm; 170-330mm; 195-350mm; 250-480mm
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Telescoping Filling Tubes For Nominal Volume 0.5, 1, 2, 5, 10 mL
FEP
Dispensette™ S (all models), III, Organic, TA and seripettor™ pro
250 to 480 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.