Learn More
BRAND™ Filling Tubes for seripettor™ Bottle Top Dispensers
Polypropylene construction. Supplied in bulk.
Brand:  BRAND™ 704534
 View more versions of this product
Product Code. 10712202
  • £9.55 / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with seripettor™ bottletop dispensers
  • Polypropylene construction
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Filling Tube
Polypropylene
Seripettor™ bottletop dispenser
500 mm