Learn More
BRAND™ Filling Valve with Seal
Includes sealing washer. For use with Dispensette™ III, seripettor™ pro and Digital Burette.
Brand: BRAND™ 6697
This item is not returnable.
View return policy
Description
- For use with Dispensette™ III, seripettor™ pro and Digital Burette
- Volume range: 0.5, 1, 2, 5, 10mL
Specifications
For Burettes | |
1 Each | |
With Sealing Washer | |
Filling Valve |