Learn More
Filter aid, Celite Hyflo Super-cel|r, Thermo Scientific Chemicals
Description
Filter aid, Celite Hyflo Super-cel™ is used as a filtration agent, a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid, Celite Hyflo Super-cel« is used as a filtration agent, a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with acids.
Specifications
Odor | Odorless |
Quantity | 2500 g |
Merck Index | 14,4973 |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Filter aid, Celite Hyflo Super-cel™ |
RUO – Research Use Only