Learn More
Thermo Scientific™ Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers
Fluoroskan™, Fluoroskan™FL, and Luminoskan™ microplate fluorometers and luminometers can be equipped with a wide range of accessories, additional excitation filters, and emission filters to expand the instrument's functionality and application flexibility.
£214.00 - £966.00
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Wavelength | Price | Quantity & Availability | |||||
15863537
|
Thermo Scientific™
11272700 |
£214.00
Each |
Estimated Shipment: 10-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12433398
|
Thermo Scientific™
1423202 |
320 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443398
|
Thermo Scientific™
1423352 |
335 nm |
£966.00
Each |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463398
|
Thermo Scientific™
142340210 |
340 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12473398
|
Thermo Scientific™
142350220 |
350 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13343087
|
Thermo Scientific™
1423552 |
355 nm |
£486.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12493398
|
Thermo Scientific™
1423602 |
360 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403408
|
Thermo Scientific™
142370212 |
370 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413408
|
Thermo Scientific™
1423802 |
380 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433408
|
Thermo Scientific™
142395240 |
395 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10221300
|
Thermo Scientific™
1424002 |
400 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13363087
|
Thermo Scientific™
1424052 |
405 nm |
£488.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443408
|
Thermo Scientific™
142405210 |
405 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11859500
|
Thermo Scientific™
1424202 |
420 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12473408
|
Thermo Scientific™
1424252 |
425 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483408
|
Thermo Scientific™
142430210 |
430 nm |
£488.00
Each |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12719516
|
Thermo Scientific™
142440210 |
440 nm |
£486.00
Each |
Estimated Shipment: 25-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403418
|
Thermo Scientific™
1424442 |
444 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413418
|
Thermo Scientific™
1424502 |
450 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12423418
|
Thermo Scientific™
1424602 |
460 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17128595
|
Thermo Scientific™
142470L |
470 nm |
£740.00
Each |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433418
|
Thermo Scientific™
142475210 |
475 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443418
|
Thermo Scientific™
1424802 |
480 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11753037
|
Thermo Scientific™
1424852 |
485 nm |
£486.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463418
|
Thermo Scientific™
142485216 |
485 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12739516
|
Thermo Scientific™
1424902 |
490 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483418
|
Thermo Scientific™
1424922 |
492 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11541559
|
Thermo Scientific™
1425102 |
510 nm |
£486.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11714958
|
Thermo Scientific™
1425152 |
515 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12493418
|
Thermo Scientific™
1425172 |
517 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11869500
|
Thermo Scientific™
1425182 |
518 nm |
£486.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403428
|
Thermo Scientific™
1425202 |
520 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413428
|
Thermo Scientific™
1425212 |
521 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17138595
|
Thermo Scientific™
142525L |
525 nm |
£740.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11834383
|
Thermo Scientific™
1425272 |
527 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11919426
|
Thermo Scientific™
1425382 |
538 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433428
|
Thermo Scientific™
142540210 |
540 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443428
|
Thermo Scientific™
1425422 |
542 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463428
|
Thermo Scientific™
142550208 |
550 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11884750
|
Thermo Scientific™
1425552 |
555 nm |
£486.00
Each |
Estimated Shipment: 17-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11894303
|
Thermo Scientific™
142560210 |
560 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12473428
|
Thermo Scientific™
142570210 |
570 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483428
|
Thermo Scientific™
1425752 |
575 nm |
£966.00
Each |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11817993
|
Thermo Scientific™
142578206 |
578 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413438
|
Thermo Scientific™
1425902 |
590 nm |
£488.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12423438
|
Thermo Scientific™
1425972 |
597 nm |
£966.00
Each |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433438
|
Thermo Scientific™
1426032 |
603 nm |
£966.00
Each |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11827993
|
Thermo Scientific™
1426042 |
604 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11741238
|
Thermo Scientific™
1426202 |
620 nm |
£488.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11731238
|
Thermo Scientific™
142620220 |
620 nm |
£966.00
Each |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12463438
|
Thermo Scientific™
1426212 |
621 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12483438
|
Thermo Scientific™
1426402 |
640 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17148595
|
Thermo Scientific™
142640L |
640 nm |
£740.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12493438
|
Thermo Scientific™
1426462 |
646 nm |
£486.00
Each |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12403448
|
Thermo Scientific™
142650220 |
650 nm |
£966.00
Each |
Estimated Shipment: 28-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12413448
|
Thermo Scientific™
1426702 |
670 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12423448
|
Thermo Scientific™
142678210 |
678 nm |
£486.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433448
|
Thermo Scientific™
1426802 |
680 nm |
£486.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12443448
|
Thermo Scientific™
1427002 |
700 nm |
£966.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12453448
|
Thermo Scientific™
1427902 |
790 nm |
£966.00
Each |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12453408
|
Thermo Scientific™
1424102 |
410 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12463408
|
Thermo Scientific™
142415210 |
415 nm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
For running fluorometric or luminometric assays, the Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers offer reliability and optical technology that has already been proven in thousands of laboratories all over the world. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility. Additionally, the Thermo Scientific™ SkanIt™ Software increases efficiency in your lab.
- Filters play an essential role in fluorometric measurements
- When ordering special filters please provide as much information as possible for the best measurement results
- If advice is needed which filters to order, contact a local Thermo Fisher Scientific representative with the following details:
- Name of the application
- Excitation and emission spectra
- Peak or center wavelengths of the excitation and emission spectra
- Fluorometric filters can also be used for luminometry depending on the application
- For luminometric applications requiring filters, contact technical support for instructions