Learn More
Spectrum Labs™ Filtration Membrane Minikap Filter
Brand: Spectrum Labs™ MK2M-204-V6N
Code : LB
Additional Details : Weight : 0.01000kg
Description
1/4inch NPT male inlet/outlet mixed cellulose ester non-sterile,225cm2 SpectrumSpecifications
0.25 in. NPT male inlet/outlet | |
MCE | |
225 cm2 |
Point of use | |
Non-sterile |