Learn More
Kartell™ Filtration and Vaccum Dessiccators
BIOPHARMA
Can be evacuated to 1bar, with loss not exceeding 0.03mbar in 24 hours. Kartell™ Filtration and Vacuum Dessicators is implosion proof.
Brand:  Kartell™ 0055400
 View more versions of this product
Product Code. 10335921
  • £633.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent polycarbonate cover seals onto a neoprene O-ring set into the flange of the polypropylene base
  • Initial vacuum established with a little grease spread on the O-ring
  • Removable pan for holding a drying agent
  • Includes vacuum release tap/non-return valve assembly
  • Specially grooved stopper in assembly allows air, when required, to re-enter very slowly to avoid disturbing the contents
Specifications
Specifications
Polypropylene, Polycarbonate
9.2 L
225 mm
1 Each
300 mm
250 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products