missing translation for 'onlineSavingsMsg'
Learn More

Corning™ FiltrEX™ 96-well Clear Filter Plates with 0.2 μm PVDF Membrane, Nonsterile

The Corning ™ FiltrEX™ 96 well clear filter plate with 0.2 μm hydrophilic PVDF membrane was designed with your application needs in mind.

Brand:  Corning™ 3508

Code : 66

Additional Details : Weight : 0.01123kg

Product Code. 10452012

  • £1503.69 / Pack of 50
Estimated Shipment: 06-12-2024
to see stock.
Add to basket

Description

Description

Specific design features include:
  • Meets the ANSI/SBS1-2004 through ANSI/SBS 4-2004 standard dimensions to ensure automation compatibility
  • Rigid sidewalls make the plate ideal for automation handling
  • Wide skirt easily accommodates bar codes
  • Individual filter discs are sealed in the wells by a proprietary over-molding process to ensure 100% integrity of each well
  • The design of the nozzle prevents cross-contamination while allowing the plate to rest on the lab bench
  • 350 μL working volume
  • Nonsterile
Specifications

Specifications

ANSI/SBS1-2004 through ANSI/SBS 4-2004
Hydrophilic PVDF
Clear
350 μL
10/Pk., 50/Cs.
Without Lid
0.2 μm
96
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers