missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Lab Markers

Product Code. p-8000127 Shop All Cole-Parmer Products
Click to view available options
Color:
Blue
Red
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11700239

Brand: Cole-Parmer™ 0996410

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Write on a variety of surfaces with these versatile lab markers. Cole-Parmer™ Lab Markers permanently mark plastic; wash off glass, metal, or porcelain. Markers won’t bleed or smudge when subjected to water or alcohol.

  • Fine-point fiber tip
  • Quick-drying ink
  • Writes on cold, wet surfaces
  • Available in four different colors

Specifications

Color Blue
For Use With (Application) For Glass, Porcelain or Metal surfaces
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.