Learn More
Epredia™ Fine Pore Cassettes
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
Brand:  Epredia™ B851735WH
278.25 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 16364096
  • £385.00 / Pack of 1000
Explore more special offers
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Fine Pore
Loose Packed
Attached Lid, Front Hinge
White
Yes
1000/Cs.
No
Cassette