Learn More
Epredia™ Fine Pore Cassettes in Tube Packs
For use with very small biopsy tissue samples. These cassettes are compatible with all Epredia PrintMate™ Cassette Printers, have a front-edge lid hinge, and attached lids that are available in 12 color varieties.
Brand: Epredia™ B851734TN
This item is not returnable.
View return policy
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Fine Pore | |
750/Cs. | |
Yes | |
For Use With PrintMate™ Printers |
Yes | |
Attached Lid, Front Hinge | |
Tan | |
Cassette |