Learn More
WA™ Disposable Pre-Inked Fingerprint Strips
An ideal choice for officers recording elimination prints at a crime scene.
Brand: WA™ B21410-400
Code : 50
Additional Details : Weight : 1.00000kg
Description
- Strips have a thin layer of fingerprint ink sandwiched between two thin sheets of plastic
- Extremely difficult to over-ink fingers because they only have a small amount of ink spread evenly between the two thin sheets
- Pack of 400 strips
Specifications
Disposable pre-inked fingerprint strips | |
Recording elimination prints at a crime scene | |
Plastic sheets coated with ink | |
400/Pk. |