missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ Dispenser

Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.

Brand:  Thermo Scientific™ 4421150

359.28 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11478409

  • £537.00 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.

Features:

  • Excellent chemical and thermal resistance
  • Fully autoclavable†
  • Exact self-locking system in the volume setting
  • Dispensing head cap prevents dripping
  • Ergonomic design prevents work fatigue
  • Threaded suction hose for bubble-free suction

Note:

Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analog Bottletop Dispenser
Range: 2-10mL, ±60.0µL
2.0 to 10.0 mL
Individual calibration certificate with serial number
1
20µL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Finnpipette™ Dispenser > Range: 2-10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Note: Not for use on hot plates.