Learn More
Thermo Scientific™ Finnpipette™ Multistepper Pipette
Reduce the risk of error and increase productivity in immunoassay applications with this multistepper pipette, designed especially for work with microplates and featuring simultaneous 8-channel dispensing.
Brand: Thermo Scientific™ 2206590
Description
Reduce the risk of error and increase productivity in immunoassay applications with the Thermo Scientific™ Finnpipette™ Multistepper Pipette. This lightweight, easy-to-use repeater pipette provides simultaneous dispensing in eight channels. Designed especially for work with microplates, the Finnpipette Multistepper helps speed up routine laboratory procedures and is easy to use — simply attach, fill, and prime the module, then dial the selected volume.
Design
- Universal handle also fits the seven single-channel Thermo Scientific™ Finntip™ Steppers
- Lightweight construction minimizes hand fatigue during repetitive dispensing
- Separate tip ejector for safe and effortless tip disposal
- Wide finger rest minimizes operator hand strain
- Module rotates a full 360°, optimizing efficiency for both right or left-handed users
Compatible with:
Accepts Finntip Multistepper 1500μL tips. They are extra long, with more air space between the liquid and tip cone to prevent contamination.
Specifications
8-Channel Module only | |
8 | |
1250 μL |
Microplate | |
Multistepper Pipette |