missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Finnpipette™ Novus GLP Kits

Click to view available options
Volume (Metric):
1 to 1000 μL
10 to 10,000 μL
10 to 1000 μL
100 to 5000 μL
Unit Size:
Each
Beschreibung
Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ Novus GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.
Novus GLP Kit 1 includes:
- Finnpipette Novus 10-100μL
- Finnpipette Novus 100-1000μL
- Two Finnpipette Novus stands
- One rack of Finntip Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Finnpipette Novus brochure
Novus GLP Kit 2 includes:
- Finnpipette Novus 100-1000μL
- Finnpipette Novus 0.5-5mL
- Two Finnpipette Novus stands
- One rack of Finntip 5ml and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Finnpipette Novus brochure
- Pipetting Guide
Novus GLP Kit 3 includes:
- Novus pipette, 1-10μL, micro
- Novus pipette, 10-100μL
- Novus pipette, 100-1000μL
- Three Finnpipette Novus pipette stands
- One rack of Flex 10, Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Novus brochure
Novus GLP Kit 4 includes:
- Novus pipette, 10-100μL
- Novus pipette, 100-1000μL
- Novus pipette, 1-10mL
- Three Finnpipette Novus pipette stands
- One rack of Flex 200, Flex 1000 and Finntip 10ml
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Novus brochure
Spezifikation
Spezifikation
| Color | Assorted |
| Model | Novus |
| Number of Channels | 1 |
| Product Line | Novus |
| Type | GLP Kit |
| Volume (Metric) | 10 to 1000 μL |
| Includes | Novus 100 and 1000 μL single channel pipettes, two Novus stands, two sample Finntip racks |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur