Learn More
Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes
Speed up repetitive dispensing into microplates with these lightweight, multichannel pipettes, with balanced weight distribution for reduced muscle stress and strain.
£1390.00 - £1930.00
Product Code | Brand | Number of Channels | Volume (Metric) | Color | Compatible Tips | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Number of Channels | Volume (Metric) | Color | Compatible Tips | Price | Quantity & Availability | ||
11720715
|
Thermo Scientific™
46300000 |
8 | 1 to 10 μL | Pink | Flex 10, 10, 20 micro, 50 micro |
£1715.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11751485
|
Thermo Scientific™
46300100 |
12 | 1 to 10 μL | Pink | Flex 10, 10, 20 micro, 50 micro |
£1390.00
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11750715
|
Thermo Scientific™
46300200 |
8 | 5 to 50 μL | Yellow | Flex 200, 200 Ext. 250 |
£1715.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11740715
|
Thermo Scientific™
46300300 |
12 | 5 to 50 μL | Yellow | Flex 200, 200 Ext. 250 |
£1820.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10727084
|
Thermo Scientific™
46300400 |
8 | 30 to 300 μL | Orange | Flex 300, 300 |
£1715.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11730715
|
Thermo Scientific™
46300500 |
12 | 30 to 300 μL | Orange | Flex 300, 300 |
£1930.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11716924
|
Thermo Scientific™
46300700 |
16 | 5 to 50 μL | Turquoise | 50 |
£1660.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11722245
|
Thermo Scientific™
46300800 |
8 | 100 to 1200 μL | Turquoise | Flex 1200 |
£1715.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Speed up repetitive dispensing into 96- and 384-microplates with Thermo Scientific™ Finnpipette™ Novus Multichannel Pipettes. Available in a wide range of models for common laboratory tasks, this product has a lightweight handle and unique index-finger operation that enable longer work periods without fatigue. Simplicity, comfort and functionality — the Novus will be your new best friend in the lab.
Features:
- Easy-to-read and intuitive menu navigation
- Multilingual user interface options
- 10 pipetting options include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more
- 9 aspirate/dispense speeds
- Save up to 9 programs with your most common protocols
- 8 multichannel versions, including a 16-channel for 384-well microplate applications, with color coding by volume for easy recognition
- Full-size, backlit screen eliminates surrounding light reflections and improves contrast in low-light conditions
- Quick in-lab calibration for accuracy and precision
- Fully autoclavable tip cone ensures sterility and avoids cross-contamination
- Long-life battery allows approximately 4000 pipetting operations; recharged in about one hour
Ergonomic Design
- Extremely lightweight construction
- Trigger-action pipetting — index finger controls dispensing with just a soft press of a button
- 120° adjustable finger rest for balance and comfort
- Soft-touch tip ejection minimizes thumb strain; can be operated by either hand
Compatible with:
Ideal for use with Thermo Scientific Finntip™ Flex or Finntip pipette tips
Includes:
Finnpipette Novus, universal charger, piston removal tool, instructions for use and calibration certificate
Specifications
Yes |