Learn More
Thermo Scientific™ Finnpipette™ Stands
GREENER_CHOICE
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
Brand:  Thermo Scientific™ 2206040
10.71 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11769855
  • £19.10 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
  • Accommodates six pipettes
  • Sturdy crossbar structure with slot for each pipette for secure storage
  • Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
  • Multi-position stand
  • Can hold any multichannel Finnpipette
  • Pipette can be stored in upright or horizontal position
  • Small footprint
Finnpipette Carousel Stand
  • Rotating, portable
  • Transparent lid protects pipettes from contamination
  • Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
  • For convenient storage of electronic Finnpipette Novus pipette
  • Accommodates both single and multichannel Finnpipette Novus models with ease
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shelf Hanger
Manual Pipette
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Finnpipette™ Stands >