Learn More
Thermo Scientific™ Finnpipette™ Stands
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
Brand: Thermo Scientific™ 9420390
27.23 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 50
Additional Details : Weight : 0.15000kg
Description
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
- Accommodates six pipettes
- Sturdy crossbar structure with slot for each pipette for secure storage
- Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
- Multi-position stand
- Can hold any multichannel Finnpipette
- Pipette can be stored in upright or horizontal position
- Small footprint
Finnpipette Carousel Stand
- Rotating, portable
- Transparent lid protects pipettes from contamination
- Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
- For convenient storage of electronic Finnpipette Novus pipette
- Accommodates both single and multichannel Finnpipette Novus models with ease
Specifications
Pipette Stand | |
Multichannel Single Pipette Stand | |
Multichannel Manual Pipette (1) | |
1 |