missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finntip™ Filtered Pipette Tips

Protect pipettes and the samples from contamination with these filtered pipette tips.

Brand:  Thermo Scientific™ 94052300

187.69 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.



Code : 04

Additional Details : Weight : 1.27000kg

 View more versions of this product

Product Code. 11782584

  • £257.00 / Pack of 960
In Stock
to see stock.
Add to basket

Description

Description

Protect pipettes and the samples from contamination with Thermo Scientific™ Finntip™ Filtered Pipette Tips. These sterile, disposable pipette tips keep pipettes clean, and are perfect for PCR amplification applications and for pipetting radioactive, infectious, and aerosol-emitting samples. Available in racks with a wide volume range of 0.2μL-10mL.

Features:

  • Inert and nonsealing polyethylene filter
  • Tips come vacuum sealed in irradiation-sterilized hinged-tip racks
  • Not autoclavable
  • Certified to be free from DNA, DNase, RNase and endotoxin
Specifications

Specifications

Finntip Filter 200µL, sterile
Filtered
Not Autoclavable
Rack
Single-use
FinnTip Filtered
200 μL
Sterile
5.2cm
Yellow
Thermo Scientific Finnpipette
Sterile & PCR Ready
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers