missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finntip™ Extended Length Pipette Tips
GREENER_CHOICE

Product Code. p-8322146
Click to view available options
Volume (Metric):
200 μL
1000 μL
Format:
Box
Bulk
Rack
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 400
Pack of 480
Pack of 960
This item is not returnable. View return policy

Product Code. 11762534

Brand: Thermo Scientific™ 9400100

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Ensure accurate, precise pipetting. Thermo Scientific™ Finntip™ Pipette Tips are manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips, to prevent liquid retention.

Key Features:

  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked or bulk

Proven quality

  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Tip Range

  • Extended length tips available for volumes 100, 200 and 1000 μL allowing for better reach with specific labware consumables

Immediate traceability

  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack packaging options

Note:

Availability may vary by country.

TRUSTED_SUSTAINABILITY

Specifications

Description Finntip 200 Ext, Non-Filtered, Non-sterile,
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Model Finntip 200 Ext
Length (English) 3 in.
Length (Metric) 7.8cm
Format Box
Color Code Yellow
For Use With (Equipment) Thermo Scientific Finnpipette 200
Product Line FinnTip
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.