Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips
GREENER_CHOICE
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Brand:  Thermo Scientific™ 9404193
119.88 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11792574
  • £214.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2.5 mL
Non-filtered
ISO 9001, ISO 14001
Thermo Scientific FinnPipette Stepper
50, 100, 150, 200, 250μL
Sterile
Individually Wrapped
FinnTip