Learn More
Fipronil, Fluka™
Analytical standard, reference material
Brand: Honeywell Fluka R1602-100MG
Additional Details : CAS Number : 120068-37-3 Weight : 0.00010kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Fipronil | |
120068-37-3 | |
White/Yellow | |
437.15 | |
5.9 to 6.1 | |
230°C (Decomposition) |
Glass Bottle | |
100 mg | |
5-amino-1-(2,6-dichloro-a,a,a-trifluoro-p-tolyl)-4-[(trifluoromethyl)sulfinyl]-1H-pyrazole-3-carbonitrile | |
Analytical Standard | |
195°C to 203°C | |
Environmental Analysis |