missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Fire exit only door alarmed sign, aluminum, 200mm x 600mm
Brand: Manutan A346105
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Conforms to the health and safety (Safety Signs and Signals) Regulations 1996. Designed to BS 5499 Parts 4 & 5 and ISO 7010.- aluminum
- Dimensions (H x W) : 200mm x 600mm