Learn More
Firefly luciferase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532208
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T , 293T transfected firefly, 293T transfected Renilla Luciferase. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.Specifications
Firefly luciferase | |
Polyclonal | |
Unconjugated | |
Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase | |
Full length Firefly Luciferase recombinant protein | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Luciferase | |
Antibody | |
IgG |